Please login to your online account to display your discounted pricing
Caframo™ PTFE Stirrer Blades
For use with Caframo Stirrers
Products
Catalog Number Mfr. No. Price Quantity    
14-500-310
caframo limited
A580
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
  • Pure PTFE, totally inert and non-scratch
  • Attachment dia.: 6.5mm, 3.2mm thick
  • Weight: 0.3 oz. (7.6g)