missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Caframo™ Pitched Blade Propeller

For use with any Caframo Overhead Stirrer

Manufacturer:  Caframo™ A533

 View more versions of this product

Catalog No. 14-501-070



Description

Description

  • Electro-polished stainless steel 316L
  • 2 in. (51mm) propeller
  • Low shear, high top-to-bottom (axial) pumping capacity with high gas injection

Order Info

Shafts sold separately

Specifications

Specifications

2 in.
Caframo™ Overhead Stirrer
Stainless Steel
0.04 lb.
5.1 cm
9.5mm Bore, Set Screw
0.019 kg
Documents