Please login to your online account to display your discounted pricing

Caffeine, 99.7%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Caffeine
CAS 58-08-2
Molecular Formula C8H10N4O2
Formula Weight 194.19
MDL Number MFCD00005758
UN Number UN1544
Beilstein 17705
Merck Index 14,1636
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3921436

  SDS
alfa aesar
3921436
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.19
Melting Point 234° to 236°C
Quantity 500g
Color White
UN Number UN1544
MDL Number MFCD00005758
CAS 58-08-2
Chemical Name or Material Caffeine
Boiling Point 178°C (subl.)
Health Hazard 1 Warning
Density 1.230
Beilstein 17705
Molecular Formula C8H10N4O2
Health Hazard 2 GHS H Statement
H302;
Harmful if swallowed.
TSCA TSCA
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a;
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
Merck Index 14,1636
Physical Form Powder
Assay 99.7%
RTECS Number EV6475000
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a;
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302;
Harmful if swallowed.