Learn More
Caffeine, 99%, Thermo Scientific Chemicals
$64.03 - $219.96
Chemical Identifiers
CAS | 58-08-2 |
---|---|
Molecular Formula | C8H10N4O2 |
Molecular Weight (g/mol) | 194.19 |
MDL Number | MFCD00005758 |
InChI Key | RYYVLZVUVIJVGH-UHFFFAOYSA-N |
Synonym | caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep |
PubChem CID | 2519 |
ChEBI | CHEBI:27732 |
SMILES | CN1C=NC2=C1C(=O)N(C)C(=O)N2C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1043122
|
Thermo Scientific Chemicals
A1043122 |
100 g |
Each for $64.03
|
|
|||||
AAA1043130
|
Thermo Scientific Chemicals
A1043130 |
250 g |
Each for $73.70
|
|
|||||
AAA104310B
|
Thermo Scientific Chemicals
A104310B |
1000 g |
Each for $219.96
|
|
|||||
Description
This material is traded for research purposes ONLY. Thermo Fisher does NOT certify this material as suitable for pharmaceutical, cosmetic, veterinary or food use. Caffeine has inhibitory action against lung and colon tumorigenses and virus induction.Caffeine is used to treat and prevent bronchopulmonary dysplasia in premature infants. It is also used for the treatment of apnea of prematurity and orthostatic hypotension. It is widely used as an additive in food, cosmetics, pharmaceuticals and beverages. It confers a protective effect against some diseases including Parkinson's disease and certain types of cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in pyrrole, tetrahydrofuran+ water mixture and ethyl acetate. Slightly soluble in water and ethanol.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
58-08-2 | |
194.19 | |
RYYVLZVUVIJVGH-UHFFFAOYSA-N | |
2519 | |
CN1C=NC2=C1C(=O)N(C)C(=O)N2C |
C8H10N4O2 | |
MFCD00005758 | |
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep | |
CHEBI:27732 |
Specifications
58-08-2 | |
1.23 | |
C8H10N4O2 | |
100 g | |
17705 | |
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep | |
RYYVLZVUVIJVGH-UHFFFAOYSA-N | |
1,3,7-trimethylpurine-2,6-dione | |
2519 | |
194.19 | |
Caffeine |
234°C to 236°C | |
99% | |
MFCD00005758 | |
UN1544 | |
14,1636 | |
Soluble in pyrrole,tetrahydrofuran+water mixture and ethyl acetate; Slightly soluble in water and ethanol. | |
CN1C=NC2=C1C(=O)N(C)C(=O)N2C | |
194.19 | |
CHEBI:27732 | |
99% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
EINECSNumber : 200-362-1
RTECSNumber : EV6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only