missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Caffeine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1043122

 View more versions of this product

Catalog No. AAA1043122



Description

Description

This material is traded for research purposes ONLY. Thermo Fisher does NOT certify this material as suitable for pharmaceutical, cosmetic, veterinary or food use. Caffeine has inhibitory action against lung and colon tumorigenses and virus induction.Caffeine is used to treat and prevent bronchopulmonary dysplasia in premature infants. It is also used for the treatment of apnea of prematurity and orthostatic hypotension. It is widely used as an additive in food, cosmetics, pharmaceuticals and beverages. It confers a protective effect against some diseases including Parkinson's disease and certain types of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in pyrrole, tetrahydrofuran+ water mixture and ethyl acetate. Slightly soluble in water and ethanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Caffeine
234°C to 236°C
99%
MFCD00005758
UN1544
14,1636
Soluble in pyrrole,tetrahydrofuran+water mixture and ethyl acetate; Slightly soluble in water and ethanol.
CN1C=NC2=C1C(=O)N(C)C(=O)N2C
194.19
CHEBI:27732
99%
58-08-2
1.23
C8H10N4O2
100 g
17705
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
RYYVLZVUVIJVGH-UHFFFAOYSA-N
1,3,7-trimethylpurine-2,6-dione
2519
194.19

Chemical Identifiers

58-08-2
194.19
RYYVLZVUVIJVGH-UHFFFAOYSA-N
2519
CN1C=NC2=C1C(=O)N(C)C(=O)N2C
C8H10N4O2
MFCD00005758
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
CHEBI:27732
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.

EINECSNumber : 200-362-1

RTECSNumber : EV6475000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only