Please login to your online account to display your discounted pricing

Cafestol, 98.92%, For HPLC analysis, MP Biomedicals™


Cafestol is a diterpene molecule present in coffee

Manufacturer: mp biomedicals  0219374550

 View more versions of this product

Catalog No. ICN19374550

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Cafestol
Name Note For HPLC analysis
CAS 469-83-0
Physical Form white powder
Molecular Formula C20H28O3
Formula Weight 316.4g/mol

Specifications

Chemical Name or Material Cafestol
Name Note For HPLC analysis
CAS 469-83-0
Physical Form white powder
Molecular Formula C20H28O3
Formula Weight 316.4g/mol