Please login to your online account to display your discounted pricing

Cafestol, 98.92%, For HPLC analysis, MP Biomedicals™

Cafestol is a diterpene molecule present in coffee

$509.11 - $509.11

Chemical Identifiers

Chemical Name or Material Cafestol
Name Note For HPLC analysis
CAS 469-83-0
Physical Form white powder
Molecular Formula C20H28O3
Formula Weight 316.4g/mol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19374550

 
mp biomedicals
0219374550
Each for $509.11
Description & Specifications

Specifications

Chemical Name or Material Cafestol
Name Note For HPLC analysis
CAS 469-83-0
Physical Form white powder
Molecular Formula C20H28O3
Formula Weight 316.4g/mol