missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium wire, 2.0mm (0.08 in.) dia., Puratronic™, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 11908BU

 View more versions of this product

Catalog No. AA11908BU



Specifications

Specifications

Cadmium wire
25cm
7440-43-9
Cd
00bh33gngh, cadmium and compounds, cadmium, elemental, ccris 112, colloidal, compounds, hsdb 282, kadmium, kadmium german, unii-00bh33gngh
[Cd]
112.414
CHEBI:22977
99.998%
≈27.2g/m
2.0mm (0.08 in.) dia.
(metals basis)
MFCD00010914
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
cadmium
23973
Wire, ≈27.2g/m
Puratronic™

Chemical Identifiers

7440-43-9
112.414
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
23973
cadmium
Cd
MFCD00010914
00bh33gngh, cadmium and compounds, cadmium, elemental, ccris 112, colloidal, compounds, hsdb 282, kadmium, kadmium german, unii-00bh33gngh
CHEBI:22977
[Cd]
Safety and Handling

Safety and Handling

GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.

P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c

H330-H341-H350-H361fd-H372-H500

EINECSNumber : 231-152-8

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents