missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium telluride, Black polycrystalline lump, 99.999% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1436506

 View more versions of this product

Catalog No. AA1436506



Specifications

Specifications

Cadmium telluride
99.999% (Metals basis)
MFCD00015998
RPPBZEBXAAZZJH-UHFFFAOYSA-N
tellanylidenecadmium
91501
Polycrystalline Lump
Black
1306-25-8
CdTe
cadmium telluride, cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride, cadmium telluride trace metals basis
[Cd]=[Te]
240.014
240
Odorless
5g

Chemical Identifiers

1306-25-8
240.014
RPPBZEBXAAZZJH-UHFFFAOYSA-N
91501
[Cd]=[Te]
CdTe
MFCD00015998
cadmium telluride, cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride, cadmium telluride trace metals basis
tellanylidenecadmium
Safety and Handling

Safety and Handling

H400-H410-H302-H312-H332: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 215-149-9

Documents