Learn More
Cadmium telluride, Black polycrystalline lump, 99.999% (Metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01436514
Description
Cadmium telluride finds applications as the semiconductor material in photovoltaics and infrared optical window. It is used in the production of thin film solar cells. It is doped with chlorine and used as a radiation detector for x-rays, alfa and beta particles. .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cadmium telluride | |
1306-25-8 | |
Polycrystalline Lump | |
99.999% (Metals basis) | |
MFCD00015998 | |
Insoluble in water. | |
[Cd]=[Te] | |
240.01 | |
240 | |
6.2 |
1041°C | |
Black | |
25 g | |
CdTe | |
cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basis | |
RPPBZEBXAAZZJH-UHFFFAOYSA-N | |
tellanylidenecadmium | |
91501 | |
Odorless |
Chemical Identifiers
1306-25-8 | |
240.01 | |
RPPBZEBXAAZZJH-UHFFFAOYSA-N | |
91501 | |
[Cd]=[Te] |
CdTe | |
MFCD00015998 | |
cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basis | |
tellanylidenecadmium |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P304+P340-P308+P313-P501c
H332-H350-H372
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 215-149-9
RTECSNumber : EV3330000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only