missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium telluride, 99.999% (metals basis), Alfa Aesar™

Used to make thin film solar cells.

Manufacturer:  Alfa Aesar 4545706

 View more versions of this product

Catalog No. AA4545706



Specifications

Specifications

Cadmium telluride
CdTe
UN3077
RPPBZEBXAAZZJH-UHFFFAOYSA-N
tellanylidenecadmium
91501
6mm and down Polycrystalline Pieces
Plastic bottle
1041°C
1306-25-8
MFCD00015998
cadmium telluride, cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride, cadmium telluride trace metals basis
[Cd]=[Te]
240.014
240.00
99.999%
6.2
5g

Chemical Identifiers

1306-25-8
240.014
RPPBZEBXAAZZJH-UHFFFAOYSA-N
91501
[Cd]=[Te]
CdTe
MFCD00015998
cadmium telluride, cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride, cadmium telluride trace metals basis
tellanylidenecadmium
Safety and Handling

Safety and Handling

TSCA : Yes

SDS Documents