Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium telluride, 99.999% (metals basis)

Semiconductors, phosphors

Chemical Identifiers

Chemical Name or Material Cadmium telluride
CAS 1306-25-8
Formula Weight 240
MDL Number MFCD00015998
UN Number UN3077
Merck Index 14,1629
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1436703

 
alfa aesar™
1436703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1436706

 
alfa aesar™
1436706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1436714

 
alfa aesar™
1436714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240
TSCA TSCA
Odor Odorless
Physical Form Polycrystalline Powder
Assay 99.999%
MDL Number MFCD00015998
Melting Point 1041°C
Quantity 1g
RTECS Number EV3330000
Chemical Name or Material Cadmium telluride
Mesh Size −325 Mesh
Assay Percent Notes (metals basis)
CAS 1306-25-8
UN Number UN3077
Merck Index 14,1629
Color Black
Solubility Information Insoluble in water and acids except HNO3 in which it is soluble with decomposition
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 6.20g/mL
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.