missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium telluride, 99.999% (metals basis)

Semiconductors, phosphors

Manufacturer:  Alfa Aesar™ 1436706

 View more versions of this product

Catalog No. AA1436706



Specifications

Specifications

Cadmium telluride
(metals basis)
MFCD00015998
14,1629
Insoluble in water and acids except HNO3 in which it is soluble with decomposition
[Cd]=[Te]
240.014
240
99.999%
−325 Mesh
Black
5g
1306-25-8
CdTe
UN3077
cadmium telluride, cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride, cadmium telluride trace metals basis
RPPBZEBXAAZZJH-UHFFFAOYSA-N
tellanylidenecadmium
91501
Polycrystalline Powder
6.20g/mL
Odorless
1041°C

Chemical Identifiers

1306-25-8
240.014
RPPBZEBXAAZZJH-UHFFFAOYSA-N
91501
[Cd]=[Te]
CdTe
MFCD00015998
cadmium telluride, cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride, cadmium telluride trace metals basis
tellanylidenecadmium
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : EV3330000

TSCA : TSCA

Documents