missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium sulfate octahydrate, ACS, 98.0 to 102.0%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1234014

 View more versions of this product

Catalog No. AA1234014



Specifications

Specifications

Cadmium sulfate octahydrate
98.0 to 102.0%
MFCD00151655
Highly soluble in water. Partially soluble in methanol and ethyl acetate. Insoluble in ethanol.
Crystalline
3.08 g/mL
White
25g
7790-84-3
3CdSO4·8H2O
White crystalline
769.50 (208.46 Anhydrous)
ACS
1.565
>40°C to H2O

Chemical Identifiers

7790-84-3
3CdSO4·8H2O
MFCD00151655
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c

H301-H330-H340-H350-H360FD-H372

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 233-331-6

RTECSNumber : EV2850000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents