Please login to your online account to display your discounted pricing

Cadmium sulfate octahydrate, ACS, 98.0-102.0%, Alfa Aesar™

Used in Standard Weston Cell, as electrolytes in electroplating, in phosphors, in manufacture of standard cadmium elements, and as a catalyst in Marsh test for AS

Chemical Identifiers

Chemical Name or Material Cadmium sulfate octahydrate
CAS 7790-84-3
Purity Grade ACS Grade
Molecular Formula 3CdSO4·8H2O
Formula Weight 769.50 (208.46 Anhydrous)
MDL Number MFCD00151655
UN Number UN2570
Merck Index 14,1627
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1234030

 
alfa aesar
1234030
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 769.50 (208.46 Anhydrous)
Melting Point >40°C (in -H2O)
Quantity 250g
Chemical Name or Material Cadmium sulfate octahydrate
Odor Odorless
Refractive Index 1.565
Assay 98.0 to 102.0%
Molecular Formula 3CdSO4·8H2O
CAS 7790-84-3
UN Number UN2570
Merck Index 14,1627
Color White
Solubility Information Soluble in water. Almost insoluble in alcohol and ethyl acetate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H330-H340-H350-H360-H372;
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 3.08
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
RTECS Number EV2850000
TSCA TSCA
Physical Form Crystalline
MDL Number MFCD00151655
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H330-H340-H350-H360-H372;
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.