missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium sulfate octahydrate, ACS, 98.0 to 102.0%, Alfa Aesar™

Manufacturer:  Alfa Aesar 12340A1*

 View more versions of this product

Catalog No. AA12340A1



Specifications

Specifications

Cadmium sulfate octahydrate
98.0 to 102.0%
MFCD00151655
769.5
ACS
1kg
7790-84-3
3CdSO4·8H2O
White crystalline
Crystalline
White

Chemical Identifiers

7790-84-3
3CdSO4·8H2O
MFCD00151655
Safety and Handling

Safety and Handling

H301-H330-H340-H350-H360-H372-H400-H410

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 233-331-6

Documents