missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Cadmium Sulfate 8/3-Hydrate, puriss. p.a. ACS Reagent, ≥99.0%, Honeywell™ Fluka™

Puriss. p.a., ACS Reagent,≥99.0% (calc. based on CdSO4 · 8/3H2O, KT)

$95.45 - $544.51

Chemical Identifiers

CAS 7790-84-3
MDL Number MFCD02253000
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045183 View Documents Honeywell Fluka™
20920100G
100g Poly Bottle Each for $251.65
 
60045187 View Documents Honeywell Fluka™
2092025G
25g Poly Bottle Each for $95.45
 
60045188 View Documents Honeywell Fluka™
20920500G
500g Poly Bottle Each for $544.51
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7790-84-3
MFCD02253000

Specifications

7790-84-3
MFCD02253000
256.57g/mol
ACS/puriss. p.a.
Poly Bottle
Colorless
100g
CdSO4 · 8/3H2O
UN2570
≥99.0% (calc. based on CdSO4 · 8/3 H2O, KT)
3.09g/cu.cm
Solid
41.5°C
Cadmium Sulfate 8/3-Hydrate
Safety and Handling

Safety and Handling

P201-P260-P301 + P310 + P330-P304 + P340 + P310-P308 + P313-P403 + P233

H301-H330-H340-H350-H360FD-H372-H410

DOTInformation : class 6.1 - PG 3 - EHS - Cadmium compounds, HI: all (not BR)

EINECSNumber : 233-331-6