Please login to your online account to display your discounted pricing

Cadmium Sulfate 8/3 Hydrate (Crystalline/Certified ACS), Fisher Chemical

$375.54 - $375.54

Chemical Identifiers

Chemical Name or Material Cadmium Sulfate 8/3 Hydrate
CAS 7790-84-3
Purity Grade Certified ACS Grade
Physical Form Solid
Assay 98.0 to 102.0%
Assay Percent Range 98.0 to 102.0 %
Molecular Formula Cd3O12S3 · 8 H2O
Formula Weight 256.52
MDL Number MFCD00151655
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

C19-500

  SDS
fisher scientific company
FLC19500
Glass Bottle 500g Each for $375.54
Case of 6 Each for $2,253.24
Description & Specifications

Specifications

Chemical Name or Material Cadmium Sulfate 8/3 Hydrate
CAS 7790-84-3
ChemAlert Storage Symbol Blue
DOT Information DOT Class 6.1, : Poison
Purity Grade Certified ACS Grade
Physical Form Solid
Assay 98.0 to 102.0%
Assay Percent Range 98.0 to 102.0 %
Molecular Formula Cd3O12S3 · 8 H2O
Formula Weight 256.52
MDL Number MFCD00151655
Calcium (Ca) 0.005% max.
Chloride 0.001% max.
Copper (Cu) 0.002% max.
Identification Pass Test
Insoluble Matter 0.005% max.
Iron (Fe) 0.001% max.
Lead (Pb) 0.003% max.
Nitrate and Nitrite 0.003% max.
Packaging Glass Bottle
Potassium (K) 0.01% max.
Sodium (Na) 0.02% max.
Zinc (Zn) 0.05% max.
Color White
Melting Point 41°C
pH 4.5 to 6.0
Quantity 500g
Safety and Handling