Please login to your online account to display your discounted pricing

Cadmium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$126.80 - $244.17

Chemical Identifiers

Chemical Name or Material Cadmium Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
View More Specs
Includes:  NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CLCD2-2Y

 
spex certiprep llc
CLCD22Y
Triple-leached LDPE bottle 125mL Each for $244.17

CLCD22M

 
spex certiprep llc
SPXCLCD22M
Triple-leached LDPE bottle 30mL Each for $126.80
Description & Specifications

Specifications

Recommended Storage Ambient
Quantity 125mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Health Hazard 1 Causes skin irritation, serious eye irritation
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
DOT Information Hazardous
Solvent or Matrix 2% HNO3
Purity Grade Trace ICP/ICP-MS Grade
Certificate of Analysis Claritas PPT™ Certificate of Analysis reports up to 68 elements at ppt levels.
CAS Min % 2.0
CAS Max % 2.0
Chemical Name or Material Cadmium Standard
CAS Max % 97.9
CAS 7440-43-9
CAS Min % 0.1
CAS Max % 0.1
CAS 7697-37-2
CAS 7732-18-5
CAS Min % 97.9

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Cd1000μg/mL

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Causes skin irritation, serious eye irritation

Ambient