missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cadmium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC CLCD22Y
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Cd | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Cadmium Standard | |
Triple-leached LDPE bottle | |
0.1,2.0,97.9 | |
Claritas PPT™ Certificate of Analysis reports up to 68 elements at ppt levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00010914 | |
[Cd] | |
112.41 | |
CHEBI:22977 | |
2% HNO3 |
7440-43-9,7732-18-5,7732-18-5 | |
125 mL | |
0.1,2.0,97.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Cd | |
BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
cadmium | |
23973 | |
Trace ICP/ICP-MS |
Chemical Identifiers
7440-43-9 | |
112.41 | |
BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
CHEBI:22977 | |
[Cd] |
Cd | |
MFCD00010914 | |
23973 | |
cadmium |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only