Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium shot, 5cm (2 in.) dia., 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Cadmium shot
CAS 7440-43-9
Name Note 5cm (2 in.) dia.
MDL Number MFCD00010914
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA39731KT

  SDS
alfa aesar™
39731KT
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay 99.9%
CAS 7440-43-9
MDL Number MFCD00010914
Chemical Name or Material Cadmium shot
Weight ≈600g/shot
Quantity 1 each
Health Hazard 3 GHS P Statement
P260-P284-P304+P340-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear respiratory protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay Percent Notes (metals basis)
Health Hazard 1 Danger
Name Note 5cm (2 in.) dia.
Health Hazard 2 GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P284-P304+P340-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear respiratory protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.