missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium selenide, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4545604

 View more versions of this product

Catalog No. AA4545604



Specifications

Specifications

Cadmium selenide
(metals basis)
MFCD00010917
14,1626
AQCDIIAORKRFCD-UHFFFAOYSA-N
selanylidenecadmium
14784
191.36
99.999%
Odorless
Black to Brown-Red
2g
1306-24-7
CdSe
UN3077
cadmium selenide, cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene
[Se]=[Cd]
191.385
CHEBI:50834
Pieces
5.81g/mL
3 to 12mm
>1350°C

Chemical Identifiers

1306-24-7
191.385
AQCDIIAORKRFCD-UHFFFAOYSA-N
14784
selanylidenecadmium
CdSe
MFCD00010917
cadmium selenide, cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene
CHEBI:50834
[Se]=[Cd]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA

SDS Documents