missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium selenide, 99.995% (metals basis), Alfa Aesar™

Used as a n-type semiconductor and its nanoparticles are used in laser diodes, opto-electronic devices, nanosensing, biomedical imaging and high efficiency solar cells. Since, it is transparent to infrared light, it finds application in photoresistors and windows for instruments utilizing IR light.

Manufacturer:  Thermo Fisher Scientific 1105718

 View more versions of this product

Catalog No. AA1105718



Specifications

Specifications

Cadmium selenide
CdSe
UN3077
Insoluble in water.
[Se]=[Cd]
191.385
CHEBI:50834
Powder
5.81
5.81
50g
1306-24-7
MFCD00010917
cadmium 2+ selenide, cadmium ii selenide, cadmium selenide, cadmium selenide candot quantum dot core , 50umol/l in hexane, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene
AQCDIIAORKRFCD-UHFFFAOYSA-N
selanylidenecadmium
14784
191.36
99.995%
Plastic bottle
>1350°C

Chemical Identifiers

1306-24-7
191.385
AQCDIIAORKRFCD-UHFFFAOYSA-N
14784
selanylidenecadmium
CdSe
MFCD00010917
cadmium 2+ selenide, cadmium ii selenide, cadmium selenide, cadmium selenide candot quantum dot core , 50umol/l in hexane, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene
CHEBI:50834
[Se]=[Cd]
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350-H373

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 215-148-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents