Please login to your online account to display your discounted pricing

Cadmium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


For atomic absorption. (1mL = 1mg Cd) Solute: Cadmium Metal Solvent: Dilute Nitric Acid

Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  SC118500

 View more versions of this product

Catalog No. SC118-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Melting Point 0°C
Boiling Point 100°C
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Color Colorless
Physical Form Liquid
Vapor Pressure 14mmHg at 16°C
Viscosity [NULL]
Density 1.0g/cm³
pH 0.5
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Chemical Name or Material Cadmium, Reference Standard Solution
Concentration 990 to 1010ppm
MDL Number MFCD00010914
CAS 7440-43-9
CAS 7697-37-2
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED CADMIUM REF. STD.
CAS 7732-18-5
Name Note 1000ppm ±1%
ChemAlert Storage Symbol White

Specifications

Chemical Name or Material Cadmium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7440-43-9
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00010914
Accessories
 

SDS & Safety Documents