Please login to your online account to display your discounted pricing

Cadmium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

For atomic absorption. (1mL = 1mg Cd) Solute: Cadmium Metal Solvent: Dilute Nitric Acid

$76.06 - $102.71

Chemical Identifiers

Chemical Name or Material Cadmium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7440-43-9
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00010914
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SC118-100

  SDS
fisher scientific company
FLSC118100
Poly Bottle 100mL Each for $76.06
Case of 6 Each for $456.36

SC118-500

  SDS
fisher scientific company
SC118500
Poly Bottle 500mL Each for $102.71
Case of 6 Each for $616.26
Description & Specifications

Specifications

Purity Grade Certified Grade
Melting Point 0°C
Boiling Point 100°C
Quantity 100mL
DOT Information DOT Class 8, : Corrosive
Color Colorless
Physical Form Liquid
Vapor Pressure 14mmHg at 16°C
Viscosity [NULL]
Density 1.0g/cm³
pH 0.5
Name Note 1000ppm ±1%
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Packaging Poly Bottle
CAS 7732-18-5
Chemical Name or Material Cadmium, Reference Standard Solution
Concentration 990 to 1010ppm
MDL Number MFCD00010914
CAS 7440-43-9
ChemAlert Storage Symbol White
CAS 7697-37-2
Traceability to NIST LOT NUMBER USED CADMIUM REF. STD.
Safety and Handling