missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium Plasma Std. Solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 14424AE

 View more versions of this product

Catalog No. AA14424AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cadmium
Clear liquid
MFCD00010914
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
Liquid
Colorless
7697-37-2,7697-37-2,7732-18-5
HNO3
Nitric acid: 5.0%; Cadmium: 1.0%; Water: 94.0%
[N+](=O)(O)[O-]
63.012
CHEBI:48107
23 hPa (17mm Hg) at 20°C
100mL

Chemical Identifiers

7697-37-2
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
CHEBI:48107
[N+](=O)(O)[O-]
HNO3
MFCD00010914
944
nitric acid
Safety and Handling

Safety and Handling

H350-H341-H373-H314-H333-H412: May cause cancer. Suspected of causing genetic defects. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May be harmful if inhaled. Harmful to aquatic life with long lasting effects.

DOTInformation : Hazard Class: 8, Packing Group: III

Documents