missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cadmium Plasma Std. Solution, Thermo Scientific Chemicals, Specpure™
Supplier: Thermo Scientific Chemicals 014424AP
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cadmium | |
500 mL | |
HNO3 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
Nitric acid: 5.0%; Cadmium: 1.0%; Water: 94.0% | |
Liquid |
7697-37-2,7732-18-5 | |
Colorless | |
MFCD00011349 | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
23 hPa (17mm Hg) at 20°C |
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
CHEBI:48107 | |
O[N+]([O-])=O |
HNO3 | |
MFCD00011349 | |
944 | |
nitric acid |
Safety and Handling
P201-P202-P234-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335-H350
DOTInformation : Hazard Class: 8, Packing Group: III
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only