missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium, plasma standard solution, Cd 1000μg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13813AP

 View more versions of this product

Catalog No. AA13813AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cadmium
Clear liquid
MFCD00010914
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
cadmium
23973
Liquid
Colorless
7440-43-9,7732-18-5,7732-18-5
Cd
Nitric acid: 5.0%; Cadmium: 0.1%; Water: 94.9%
[Cd]
112.414
CHEBI:22977
23 hPa (17mm Hg) at 20°C
500mL

Chemical Identifiers

7440-43-9
112.414
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
CHEBI:22977
[Cd]
Cd
MFCD00010914
23973
cadmium
Safety and Handling

Safety and Handling

When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

H350-H314-H333: May cause cancer. Causes severe skin burns and eye damage. May be harmful if inhaled.

DOTInformation : Hazard Class: 8, Packing Group: III

Documents