Learn More
Cadmium phosphate (ortho), 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04020118
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cadmium phosphate (ortho) | |
13847-17-1 | |
50 g | |
MFCD00049414 | |
cadmium phosphate, cadmium orthophosphate, tricadmium diphosphate, cadmium ii phosphate, tricadmium bis phosphate, cadmium phosphte cd3 po4 2, cadmium phosphate ortho, phosphoric acid, cadmium salt 2:3, acmc-20alja, cadmium 2+ diphosphate | |
NRGIRRZWCDKDMV-UHFFFAOYSA-H | |
cadmium(2+);diphosphate | |
26060 | |
99% |
∼1500°C | |
Powder | |
Cd3O8P2 | |
UN2570 | |
Insoluble in water | |
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cd+2].[Cd+2].[Cd+2] | |
527.182 | |
527.14 |
Chemical Identifiers
13847-17-1 | |
527.182 | |
NRGIRRZWCDKDMV-UHFFFAOYSA-H | |
26060 | |
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cd+2].[Cd+2].[Cd+2] |
Cd3O8P2 | |
MFCD00049414 | |
cadmium phosphate, cadmium orthophosphate, tricadmium diphosphate, cadmium ii phosphate, tricadmium bis phosphate, cadmium phosphte cd3 po4 2, cadmium phosphate ortho, phosphoric acid, cadmium salt 2:3, acmc-20alja, cadmium 2+ diphosphate | |
cadmium(2+);diphosphate |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber : 237-581-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only