missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium oxide, 99.998% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1066436

 View more versions of this product

Catalog No. AA1066436



Specifications

Specifications

Cadmium oxide
99.99% (metals basis)
MFCD00010921
CXKCTMHTOKXKQT-UHFFFAOYSA-N
oxocadmium
14782
8.15
1306-19-0
CdO
aska-rid
O=[Cd]
128.413
Powder
500g

Chemical Identifiers

1306-19-0
128.413
CXKCTMHTOKXKQT-UHFFFAOYSA-N
14782
O=[Cd]
CdO
MFCD00010921
aska-rid
oxocadmium
Safety and Handling

Safety and Handling

EINECSNumber : 215-146-2

Documents