Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium, Oil based standard solution, Specpure™, Cd 1000μg/g

Chemical Identifiers

Chemical Name or Material Cadmium Oil based standard solution
Name Note Cd 1000μg/g
Purity Grade Specpure™
MDL Number MFCD00010914
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1422818

  SDS
alfa aesar™
1422818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 1 Danger
Purity Grade Specpure™
Physical Form Liquid
Quantity 50g
Name Note Cd 1000μg/g
MDL Number MFCD00010914
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Chemical Name or Material Cadmium Oil based standard solution
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.