Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium iodide, ultra dry, 99.9985% (metals basis)

Chemical Identifiers

Chemical Name or Material Cadmium iodide
CAS 7790-80-9
Name Note Ultra dry
Formula Weight 366.21
MDL Number MFCD00010920
UN Number UN2570
Merck Index 14,1621
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3568303

  SDS
alfa aesar™
3568303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3568306

  SDS
alfa aesar™
3568306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3568314

  SDS
alfa aesar™
3568314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 366.21
Packaging Ampule
Color White to yellow
Sensitivity Light Sensitive & Hygroscopic
MDL Number MFCD00010920
Health Hazard 1 Danger
Density 5.67g/mL
TSCA TSCA
Mesh Size −10 Mesh
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
Storage Note 1
  • Ampouled under argon
UN Number UN2570
Physical Form Beads
Chemical Name or Material Cadmium iodide
Odor Odorless
RTECS Number EV1290000
Name Note Ultra dry
Boiling Point 787°C
Merck Index 14,1621
Assay Percent Notes (metals basis)
Quantity 1g
Melting Point 387° to 389°C
CAS 7790-80-9
Health Hazard 2 GHS H Statement
H301-H331-H351-H373
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P261-P281-P301+P310-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99.9985%

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P301+P310-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H351-H373
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.

  • Ampouled under argon