missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium iodide, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 1312522

 View more versions of this product

Catalog No. AA1312522



Description

Description

  • In photography, process engraving and lithography, analytical chemistry, electrodeposition of Cadmium, manufacturing of phosphors, lubricants
Specifications

Specifications

Cadmium iodide
(metals basis)
MFCD00010920
14,1621
Soluble in water, alcohol, ether, acetone, ammonia, and acids
[Cd+2].[I-].[I-]
366.223
366.21
5.67g/mL
Odorless
787°C
387°C to 389°C
7790-80-9
CdI2
UN2570
cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g
OKIIEJOIXGHUKX-UHFFFAOYSA-L
cadmium(2+);diiodide
24635
99.5%
−40 Mesh
Light Sensitive & Hygroscopic
Green-Yellow
100g

Chemical Identifiers

7790-80-9
366.223
OKIIEJOIXGHUKX-UHFFFAOYSA-L
24635
[Cd+2].[I-].[I-]
CdI2
MFCD00010920
cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g
cadmium(2+);diiodide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H351-H373
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P281-P301+P310-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS

RTECSNumber : EV1290000

TSCA : TSCA

Documents