Please login to your online account to display your discounted pricing

Cadmium ICP Standard, 1mL = 1mg Cd (1,000ppm Cd), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Cadmium ICP Standard
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
CAS Min % 4.56
CAS Max % 4.74
CAS 7440-43-9
CAS Min % 0.09
CAS Max % 0.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PCD1KN-100

 
ricca chemical company
RIPCD1KN100
Poly natural 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PCD1KN-500

 
ricca chemical company
RIPCD1KN500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 100mL
Chemical Name or Material Cadmium ICP Standard
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
Packaging Poly natural
CAS Min % 4.56
Concentration or Composition Notes 1,000ppm Cd
Equivalence 1 mL = 1 mg Cd
CAS Max % 0.1
CAS 7732-18-5
CAS Max % 4.74
CAS 7440-43-9
CAS Min % 0.09