missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cadmium ICP Standard, 10,000 ppm Cd in 5% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PCD10KN100
Specifications
Cadmium ICP Standard | |
Natural LDPE Bottle | |
87.0,9.84,0.87 | |
Colorless | |
Cd | |
WLZRMCYVCSSEQC-UHFFFAOYSA-N | |
cadmium(2+) ion | |
23973 | |
1 mL = 10 mg |
7440-43-9,7697-37-2,7440-43-9 | |
100 mL | |
91.0,10.24,0.91 | |
10,000ppm Cd | |
MFCD00010914 | |
[Cd++] | |
112.41 | |
CHEBI:22977 |