missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium hydroxide, Cd 75% min

Cadmium plating, storage battery electrodes

Manufacturer:  Alfa Aesar™ 8929736

 View more versions of this product

Catalog No. AA8929736



Specifications

Specifications

Cadmium hydroxide
Cd(OH)2
UN2570
Soluble in dilute acids, NH4Cl solutions
Crystalline
4.79g/mL at 15°C
300°C (decomposition)
21041-95-2
MFCD00015995
14,1620
146.41
Cd 75% min
Air Sensitive
500g

Chemical Identifiers

21041-95-2
Cd(OH)2
MFCD00015995
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS

RTECSNumber : EV1260000

TSCA : TSCA

SDS Documents