missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium foil, 0.5mm (0.02 in.) thick, 99.85% (metals basis)

Manufacturer:  Alfa Aesar™ 10121GX

 View more versions of this product

Catalog No. AA10121GX



Specifications

Specifications

Cadmium foil
200 x 300mm
7440-43-9
Cd
00bh33gngh
[Cd]
112.414
CHEBI:22977
99.85%
≈65g/100x150mm
0.5mm (0.02 in.) thick
(metals basis)
MFCD00010914
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
cadmium
23973
Foil

Chemical Identifiers

7440-43-9
112.414
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
23973
cadmium
Cd
MFCD00010914
00bh33gngh
CHEBI:22977
[Cd]
Safety and Handling

Safety and Handling

GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.

GHS P Statement
P260-P284-P304+P340-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear respiratory protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents