Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium cyclohexanebutyrate

As an organophile Cadmium compound

Chemical Identifiers

Chemical Name or Material Cadmium cyclohexanebutyrate
CAS 55700-14-6
Formula Weight 450.9
MDL Number MFCD00036398
UN Number UN2570
Synonym Cyclohexanebutyric acid cadmium salt; 4-Cyclohexylbutyric acid cadmium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4197003

 
alfa aesar™
4197003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 450.9
Chemical Name or Material Cadmium cyclohexanebutyrate
TSCA TSCA
Physical Form Powder
MDL Number MFCD00036398
CAS 55700-14-6
UN Number UN2570
Synonym Cyclohexanebutyric acid cadmium salt; 4-Cyclohexylbutyric acid cadmium salt
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.