Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium, course powder, 1-4mm for reductors, ≥99% (metals basis)

$75.19 - $75.19

Chemical Identifiers

Chemical Name or Material Cadmium
CAS 7440-43-9
Name Note 1 to 4mm for reductors
MDL Number MFCD00010914
UN Number UN2570
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4724614

  SDS
alfa aesar™
4724614
25g Each for $75.19

AA4724622

  SDS
alfa aesar™
4724622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4724636

  SDS
alfa aesar™
4724636
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Coarse Powder
Health Hazard 2 GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
CAS 7440-43-9
TSCA TSCA
Name Note 1 to 4mm for reductors
Quantity 25g
Assay Percent Notes (metals basis)
Chemical Name or Material Cadmium
Health Hazard 1 Danger
UN Number UN2570
RTECS Number EU9800000
Assay ≥99%
MDL Number MFCD00010914
Health Hazard 3 GHS P Statement
P260-P284-P304+P340-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear respiratory protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: CADMIUM COMPOUNDS
Safety and Handling

Danger

GHS P Statement
P260-P284-P304+P340-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear respiratory protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.