missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium chloride, White powder, 99.99% (Metals basis), Alfa Aesar™

$138.01 - $376.28

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.314
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym caddy
PubChem CID 24947
ChEBI CHEBI:35456
IUPAC Name dichlorocadmium
SMILES Cl[Cd]Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1186014 View Documents Alfa Aesar
1186014
25g Each for $138.01
 
AA1186022 View Documents Alfa Aesar
1186022
100g Each for $376.28
 
Specifications

Chemical Identifiers

10108-64-2
183.314
YKYOUMDCQGMQQO-UHFFFAOYSA-L
24947
dichlorocadmium
CdCl2
MFCD00010916
caddy
CHEBI:35456
Cl[Cd]Cl

Specifications

10108-64-2
CdCl2
caddy
Cl[Cd]Cl
183.314
CHEBI:35456
Powder
Odorless
White
Cadmium chloride
99.99% (Metals basis)
MFCD00010916
YKYOUMDCQGMQQO-UHFFFAOYSA-L
dichlorocadmium
24947
183.32
4.047g/cm3
Anhydrous
25g
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

H301-H330-H340-H350-H360-H372-H400-H410: Toxic if swallowed. Fatal if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 233-296-7