missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium chloride, White powder, 99.99% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1186022

 View more versions of this product

Catalog No. AA1186022



Specifications

Specifications

Cadmium chloride
99.99% (Metals basis)
MFCD00010916
YKYOUMDCQGMQQO-UHFFFAOYSA-L
dichlorocadmium
24947
183.32
Odorless
White
10108-64-2
CdCl2
caddy
Cl[Cd]Cl
183.314
CHEBI:35456
Powder
Anhydrous
100g

Chemical Identifiers

10108-64-2
183.314
YKYOUMDCQGMQQO-UHFFFAOYSA-L
24947
dichlorocadmium
CdCl2
MFCD00010916
caddy
CHEBI:35456
Cl[Cd]Cl
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

H301-H330-H340-H350-H360-H372-H400-H410: Toxic if swallowed. Fatal if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 233-296-7

Documents