missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium chloride, ultra dry, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 1366714

 View more versions of this product

Catalog No. AA1366714



Description

Description

  • Ampouled under argon
Specifications

Specifications

Cadmium chloride
10108-64-2
CdCl2
UN2570
cadmium chloride, cadmium dichloride, caddy, vi-cad, cadmium chloride, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
Cl[Cd]Cl
183.314
CHEBI:35456
Beads
4.047g/mL
Odorless
Hygroscopic
568°C
Ultra dry
(metals basis)
MFCD00010916
14,1617
YKYOUMDCQGMQQO-UHFFFAOYSA-L
dichlorocadmium
24947
183.32
99.998%
−10 Mesh
Ampule
960°C
25g

Chemical Identifiers

10108-64-2
183.314
YKYOUMDCQGMQQO-UHFFFAOYSA-L
24947
dichlorocadmium
CdCl2
MFCD00010916
cadmium chloride, cadmium dichloride, caddy, vi-cad, cadmium chloride, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
CHEBI:35456
Cl[Cd]Cl
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H340-H350-H360-H372
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS

RTECSNumber : EV0175000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Documents