Please login to your online account to display your discounted pricing

Cadmium chloride, ultra dry, 99.996% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Cadmium chloride
CAS 10108-64-2
Physical Form Powder
Assay Percent Range 99.996% (Metal basis)
Molecular Formula CdCl2
Formula Weight 183.32
MDL Number MFCD00010916
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4285204

 
alfa aesar
4285204
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4285209

 
alfa aesar
4285209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4285218

 
alfa aesar
4285218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4285230

 
alfa aesar
4285230
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 183.32
Quantity 2g
Color White
DOT Information DOT Class: 6.1, Packaging Group: III
Chemical Name or Material Cadmium chloride
EINECS Number 233-296-7
Odor Odorless
Physical Form Powder
Assay Percent Range 99.996% (Metal basis)
Molecular Formula CdCl2
Appearance White Various forms (powder/flake/crystalline/beads
MDL Number MFCD00010916
CAS 10108-64-2
Health Hazard 1 H301-H330-H340-H350-H360-H372-H400-H410 : Toxic if swallowed. Fatal if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
Density 4.047g/cm3 at 20°C
Safety and Handling

H301-H330-H340-H350-H360-H372-H400-H410 : Toxic if swallowed. Fatal if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.