Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium chloride, ultra dry, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Cadmium chloride
CAS 10108-64-2
Name Note Ultra dry
Formula Weight 183.32
MDL Number MFCD00010916
UN Number UN2570
Merck Index 14,1617
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3572809

  SDS
alfa aesar™
3572809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3572818

  SDS
alfa aesar™
3572818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 183.32
Packaging Ampule
Chemical Name or Material Cadmium chloride
TSCA TSCA
Mesh Size −10 Mesh
Odor Odorless
Name Note Ultra dry
Physical Form Beads
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00010916
CAS 10108-64-2
UN Number UN2570
Merck Index 14,1617
Boiling Point 960°C
Melting Point 568°C
Quantity 10g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H330-H340-H350-H360-H372
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
Density 4.047g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
RTECS Number EV0175000

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H330-H340-H350-H360-H372
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.

  • Ampouled under argon