missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01066118

 View more versions of this product

Catalog No. AA1066118



Description

Description

Cadmium chloride hydrate is used in dyeing, printing, textiles and in metal finishing baths. It is also used as a reagent to make photographic paper and carbon paper. Further, it is utilized as an absorbing agent to absorb hydrogen sulfide as well as analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cadmium chloride hydrate
Crystalline Powder
(metals basis)
MFCD00149627
14,1617
Soluble in water and acetone. Slightly soluble in methanol. Insoluble in ether.
[Cl-].[Cl-].[Cd++]
183.31
183.32 (Anhydrous)
Puratronic™
Hygroscopic
654054-66-7
50 g
CdCl2
UN2570
cadmium chloride, ultra dry, cdcl2.h2o, cadmium chloride hydrate, cadmium chloride 1-hydrate, cadmium chloride hydrate, puratronic, cadmium chloride monohydrate, puriss, cadmium chloride hydrate trace metals basis
YKYOUMDCQGMQQO-UHFFFAOYSA-L
dichlorocadmium;hydrate
16211517
99.998%
1.6513

Chemical Identifiers

654054-66-7
183.31
YKYOUMDCQGMQQO-UHFFFAOYSA-L
16211517
CdCl2
MFCD00149627
cadmium chloride, ultra dry, cdcl2.h2o, cadmium chloride hydrate, cadmium chloride 1-hydrate, cadmium chloride hydrate, puratronic, cadmium chloride monohydrate, puriss, cadmium chloride hydrate trace metals basis
[Cl-].[Cl-].[Cd++]
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H340-H350-H360-H372
Fatal if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c

H301-H330-H340-H350-H360-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CADMIUM COMPOUNDS

EINECSNumber : 233-296-7

RTECSNumber : EV0175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only