Please login to your online account to display your discounted pricing

Cadmium Chloride Hemipentahydrate (Crystalline/Certified ACS), Fisher Chemical


Quantity 500g
Packaging Glass Bottle

Manufacturer: fisher scientific company  FLC9500

 View more versions of this product

Catalog No. C9-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified ACS Grade
Physical Form Crystalline Solid
Formula Weight 228.35
Melting Point 568°C
Quantity 500g
DOT Information DOT Class 6.1, : Poison
Assay 79.5 to 81.0%
Molecular Formula CdCl2 · 2.5 H2O
Vapor Pressure 10mmHg at 656°C
Viscosity [NULL]
Anhydrous or Hydrate Hemipentahydrate
Color White
Identification Pass Test
Lead (Pb) 0.005% max.
Packaging Glass Bottle
Potassium (K) 0.02% max.
Assay Percent Range 79.5 to 81.0 %
Sodium (Na) 0.05% max.
CAS 7790-78-5
Copper (Cu) 5ppm max.
Chemical Name or Material Cadmium Chloride
Iron (Fe) 5ppm max.
Nitrate and Nitrite 0.003% max.
Ammonium 0.005% max.
Sulfate 0.005% max.
MDL Number MFCD00799071
ChemAlert Storage Symbol Blue
Zinc (Zn) 0.05% max.
Calcium (Ca) 0.005% max.

Specifications

Chemical Name or Material Cadmium Chloride
CAS 7790-78-5
Purity Grade Certified ACS Grade
Physical Form Crystalline Solid
Assay 79.5 to 81.0%
Assay Percent Range 79.5 to 81.0 %
Molecular Formula CdCl2 · 2.5 H2O
Formula Weight 228.35
MDL Number MFCD00799071
Accessories
 

SDS & Safety Documents