Please login to your online account to display your discounted pricing

Cadmium Chloride Hemipentahydrate (Crystalline/Certified ACS), Fisher Chemical

$367.08 - $367.08

Chemical Identifiers

Chemical Name or Material Cadmium Chloride
CAS 7790-78-5
Purity Grade Certified ACS Grade
Physical Form Crystalline Solid
Assay 79.5 to 81.0%
Assay Percent Range 79.5 to 81.0 %
Molecular Formula CdCl2 · 2.5 H2O
Formula Weight 228.35
MDL Number MFCD00799071
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

C9-500

  SDS
fisher scientific company
FLC9500
Glass Bottle 500g Each for $367.08
Case of 6 Each for $2,202.48
Description & Specifications

Specifications

Purity Grade Certified ACS Grade
Physical Form Crystalline Solid
Formula Weight 228.35
Melting Point 568°C
Quantity 500g
DOT Information DOT Class 6.1, : Poison
Assay 79.5 to 81.0%
Color White
Vapor Pressure 10mmHg at 656°C
Molecular Formula CdCl2 · 2.5 H2O
Anhydrous or Hydrate Hemipentahydrate
Lead (Pb) 0.005% max.
Copper (Cu) 5ppm max.
Sodium (Na) 0.05% max.
Packaging Glass Bottle
Potassium (K) 0.02% max.
Identification Pass Test
CAS 7790-78-5
Assay Percent Range 79.5 to 81.0 %
MDL Number MFCD00799071
Sulfate 0.005% max.
Calcium (Ca) 0.005% max.
Nitrate and Nitrite 0.003% max.
Iron (Fe) 5ppm max.
Ammonium 0.005% max.
Zinc (Zn) 0.05% max.
ChemAlert Storage Symbol Blue
Chemical Name or Material Cadmium Chloride
Safety and Handling