Please login to your online account to display your discounted pricing

Cadmium Chloride Anhydrous (White Granular Powder/Certified ACS), Fisher Chemical


Quantity 500g
Packaging Glass Bottle

Manufacturer: fisher scientific company  FLC10500

 View more versions of this product

Catalog No. C10-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99%
Melting Point 568°C
Boiling Point 960°C
Purity Grade Certified ACS Grade
Formula Weight 183.35
Quantity 500g
DOT Information DOT Class 6.1, : Poison
Physical Form Solid
Color White
Vapor Pressure 10mmHg at 656°C
Sodium (Na) 0.05% max.
Calcium (Ca) 0.01% max.
Assay Percent Range ≥99 %
Identification Pass Test
Nitrate and Nitrite 0.003% max.
ChemAlert Storage Symbol Blue
MDL Number MFCD00010916
Copper (Cu) 0.001% max.
Packaging Glass Bottle
Lead (Pb) 0.005% max.
Insoluble Matter 0.01% max.
Sulfate 0.005% max.
Ammonium 0.01% max.
Iron (Fe) 0.001% max.
Potassium (K) 0.02% max.
Zinc (Zn) 0.05% max.
Chemical Name or Material Cadmium Chloride Anhydrous
Molecular Formula CdCl2
CAS 10108-64-2

Specifications

Chemical Name or Material Cadmium Chloride Anhydrous
CAS 10108-64-2
Purity Grade Certified ACS Grade
Physical Form Solid
Assay ≥99%
Assay Percent Range ≥99 %
Molecular Formula CdCl2
Formula Weight 183.35
MDL Number MFCD00010916
Accessories
 

SDS & Safety Documents