Please login to your online account to display your discounted pricing

Cadmium Chloride Anhydrous (White Granular Powder/Certified ACS), Fisher Chemical

$216.38 - $525.77

Chemical Identifiers

Chemical Name or Material Cadmium Chloride Anhydrous
CAS 10108-64-2
Purity Grade Certified ACS Grade
Physical Form Solid
Assay ≥99%
Assay Percent Range ≥99 %
Molecular Formula CdCl2
Formula Weight 183.35
MDL Number MFCD00010916
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

C10-500

  SDS
fisher scientific company
FLC10500
Glass Bottle 500g Each for $525.77
Case of 6 Each for $3,154.62

C10-100

  SDS
fisher scientific company
FLC10100
Glass Bottle 100g Each for $216.38
Case of 6 Each for $1,298.28
Description & Specifications

Specifications

Assay ≥99%
Purity Grade Certified ACS Grade
Formula Weight 183.35
Boiling Point 960°C
Melting Point 568°C
Quantity 500g
DOT Information DOT Class 6.1, : Poison
Physical Form Solid
Vapor Pressure 10mmHg at 656°C
Viscosity [NULL]
Color White
Identification Pass Test
Nitrate and Nitrite 0.003% max.
Assay Percent Range ≥99 %
Sodium (Na) 0.05% max.
MDL Number MFCD00010916
ChemAlert Storage Symbol Blue
Calcium (Ca) 0.01% max.
Chemical Name or Material Cadmium Chloride Anhydrous
Insoluble Matter 0.01% max.
Iron (Fe) 0.001% max.
Lead (Pb) 0.005% max.
Packaging Glass Bottle
Potassium (K) 0.02% max.
Molecular Formula CdCl2
Ammonium 0.01% max.
Sulfate 0.005% max.
CAS 10108-64-2
Zinc (Zn) 0.05% max.
Copper (Cu) 0.001% max.
Safety and Handling