Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium chloride, anhydrous, 94%

Chemical Identifiers

Chemical Name or Material Cadmium chloride
CAS 10108-64-2
Name Note Anhydrous
Formula Weight 183.32g/mol
MDL Number MFCD00010916
UN Number UN2570
Merck Index 14,1617
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1251018

 
alfa aesar™
12510-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1251030

 
alfa aesar™
1251030
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 4.047g/mL
Odor Odorless
Quantity 50g
Boiling Point 960°C
Melting Point 568°C
Quantity 50g
Chemical Name or Material Cadmium chloride
Name Note Anhydrous
CAS 10108-64-2
Physical Form Powder
Assay 94%
Formula Weight 183.32g/mol
MDL Number MFCD00010916
UN Number UN2570
Merck Index 14,1617
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H330-H340-H350-H360-H372
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
RTECS Number EV0175000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H330-H340-H350-H360-H372
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.